Assay Details
Gene Symbol: Ccdc82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007660.1 | NP_001007661.1 | 3 - 4 | 974 | 70 | |
XM_006242519.2 | XP_006242581.1 | 4 - 5 | 1036 | 70 | ||
GenBank mRNA | BC081989.1 | - | 3 - 4 | 974 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
