Assay Details
Gene Symbol: Ap1m2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108996.2 | NP_001102466.1 | 9 - 10 | 1083 | 60 | |
GenBank mRNA | BC167082.1 | - | 9 - 10 | 1083 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
