Assay Details
Gene Symbol: Cdc37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053743.1 | NP_446195.1 | 3 - 4 | 579 | 103 | |
GenBank mRNA | AB097113.1 | - | 3 - 4 | 492 | 103 | |
BC061720.1 | - | 3 - 4 | 548 | 103 | ||
D26564.1 | - | 3 - 4 | 579 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
