Assay Details
Gene Symbol: Casp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053736.2 | NP_446188.2 | 4 - 5 | 779 | 80 | |
GenBank mRNA | BC104720.1 | - | 4 - 5 | 779 | 80 | |
FQ227402.1 | - | 4 - 5 | 816 | 80 | ||
FQ232279.1 | - | 4 - 5 | 816 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
