Assay Details
Gene Symbol: Slc39a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106124.1 | NP_001099594.1 | 3 - 4 | 664 | 89 | |
XM_006254285.2 | XP_006254347.1 | 5 - 6 | 1300 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
