Assay Details
Gene Symbol: Bmi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107368.2 | NP_001100838.1 | 2 - 3 | 199 | 85 | |
XM_008771871.1 | XP_008770093.1 | 2 - 3 | 3084 | 85 | ||
GenBank mRNA | BC168209.1 | - | 2 - 3 | 199 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
