Assay Details
Gene Symbol: Olah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022705.1 | NP_073196.1 | 3 - 4 | 482 | 83 | |
XM_008771886.1 | XP_008770108.1 | 4 - 5 | 636 | 83 | ||
XM_008771887.1 | XP_008770109.1 | 3 - 4 | 339 | 83 | ||
GenBank mRNA | M16200.1 | - | 3 - 4 | 482 | 83 | |
Y00311.1 | - | 2 - 3 | 217 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
