Assay Details
Gene Symbol: Rsu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109404.1 | NP_001102874.1 | 7 - 8 | 875 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P378362 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P719179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219906 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152521 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315282 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854054 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117001 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918900 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835371 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234739 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117068 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962344 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776473 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47934 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P719179 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P378362 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P719179 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P378362 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P719179 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P378362 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P378362 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 86828 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 77270 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 85135 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 64468 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 98734 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57098 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10799740 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799742 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799744 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799738 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799743 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799739 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799737 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799741 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10799736 | - | - |
Rat Genome 230 2.0 Array | 1379423_at | - | - |
Rat Genome 230 2.0 Array | 1379423_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1379423_at | - | - |
Affymetrix HT Rat Focus Array | 1379423_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1700015099.rn.1 | TC1700002251.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700015101.rn.1 | TC1700002251.rn.1 | RTA TC |
Rat Expression Set 230 | 1379423_PM_at | - | - |
Rat Expression Set 230 | 1379423_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799736 | - | - |
Rat Genome U34 Set | rc_AA963364_at | - | - |
Rat Genome U34 Set | rc_AI176004_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20997404 | - | - |
