Assay Details
Gene Symbol: Prpf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138523.2 | NP_612532.1 | 8 - 9 | 898 | 123 | |
GenBank mRNA | AF244920.1 | - | 8 - 9 | 893 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
