Assay Details
Gene Symbol: Hacd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006254293.2 | XP_006254355.1 | 3 - 4 | 1098 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
