Assay Details
Gene Symbol: Sephs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104630.1 | NP_001098100.1 | 3 - 4 | 656 | 104 | |
XM_006254266.2 | XP_006254328.1 | 3 - 4 | 2283 | 104 | ||
GenBank mRNA | AY724491.1 | - | 3 - 4 | 656 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
