Assay Details
Gene Symbol: Nmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207590.1 | NP_997473.1 | 10 - 11 | 1094 | 80 | |
XM_008771854.1 | XP_008770076.1 | 10 - 11 | 1113 | 80 | ||
XM_008771855.1 | XP_008770077.1 | 8 - 9 | 1116 | 80 | ||
XM_008771856.1 | XP_008770078.1 | 9 - 10 | 1062 | 80 | ||
GenBank mRNA | AJ629217.1 | - | 10 - 11 | 1094 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
