Assay Details
Gene Symbol: Lrp2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017443.1 | NP_001017443.1 | 7 - 8 | 991 | 149 | |
XM_006253141.2 | XP_006253203.1 | 7 - 8 | 907 | 149 | ||
XM_006253143.2 | XP_006253205.1 | 8 - 9 | 1988 | 149 | ||
XM_008771257.1 | XP_008769479.1 | 6 - 7 | 793 | 149 | ||
GenBank mRNA | BC092575.1 | - | 7 - 8 | 991 | 149 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
