Assay Details
Gene Symbol: Enpp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107311.1 | NP_001100781.1 | 1 - 2 | 307 | 82 | |
XM_006253156.2 | XP_006253218.1 | 1 - 2 | 325 | 82 | ||
XM_008771259.1 | XP_008769481.1 | 1 - 2 | 325 | 82 | ||
GenBank mRNA | BC158772.1 | - | 1 - 2 | 287 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
