Assay Details
Gene Symbol: Casp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012922.2 | NP_037054.1 | 5 - 6 | 543 | 59 | |
XM_006253130.2 | XP_006253192.1 | 5 - 6 | 587 | 59 | ||
GenBank mRNA | BC081854.1 | - | 5 - 6 | 543 | 59 | |
U49930.1 | - | 4 - 5 | 513 | 59 | ||
U84410.1 | - | 4 - 5 | 517 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
