Assay Details
Gene Symbol: Cenpu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025673.1 | NP_001020844.1 | 9 - 10 | 1082 | 139 | |
GenBank mRNA | BC097399.1 | - | 9 - 10 | 1082 | 139 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
