Assay Details
Gene Symbol: Primpol | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253167.2 | XP_006253229.1 | 4 - 5 | 458 | 119 | |
XM_341433.6 | XP_341434.3 | 4 - 5 | 455 | 119 | ||
GenBank mRNA | FQ230438.1 | - | 1 - 2 | 81 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
