Assay Details
Gene Symbol: Primpol | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253167.2 | XP_006253229.1 | 6 - 7 | 731 | 83 | |
XM_341433.6 | XP_341434.3 | 6 - 7 | 728 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
