Assay Details
Gene Symbol: Primpol | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253167.2 | XP_006253229.1 | 7 - 8 | 964 | 152 | |
XM_341433.6 | XP_341434.3 | 7 - 8 | 961 | 152 | ||
GenBank mRNA | FQ230438.1 | - | 4 - 5 | 587 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
