Assay Details
Gene Symbol: Gmpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013036.1 | NP_001013054.1 | 4 - 5 | 376 | 84 | |
XM_006251925.2 | XP_006251987.1 | 4 - 5 | 437 | 84 | ||
XM_006251926.2 | XP_006251988.1 | 4 - 5 | 437 | 84 | ||
XM_006251927.2 | XP_006251989.1 | 5 - 6 | 532 | 84 | ||
GenBank mRNA | BC089848.1 | - | 4 - 5 | 376 | 84 | |
FQ211507.1 | - | 5 - 6 | 472 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
