Assay Details
Gene Symbol: Lats2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107267.1 | NP_001100737.1 | 4 - 5 | 2516 | 69 | |
XM_008770748.1 | XP_008768970.1 | 5 - 6 | 2561 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
