Assay Details
Gene Symbol: Dhrs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001078405.6 | XP_001078405.4 | 3 - 4 | 265 | 119 | |
XM_008770715.1 | XP_008768937.1 | 3 - 4 | 281 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
