Assay Details
Gene Symbol: Slc7a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031341.1 | NP_112631.1 | 1 - 2 | 150 | 73 | |
GenBank mRNA | AB020520.1 | - | 1 - 2 | 150 | 73 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB020520.1, NM_031341.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
