Assay Details
Gene Symbol: Rcbtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108380.1 | NP_001101850.1 | 2 - 3 | 496 | 84 | |
XM_008770759.1 | XP_008768981.1 | 3 - 4 | 322 | 84 | ||
GenBank mRNA | AY724518.1 | - | 1 - 2 | 142 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
