Assay Details
Gene Symbol: Rcbtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108380.1 | NP_001101850.1 | 3 - 4 | 651 | 132 | |
GenBank mRNA | AY724518.1 | - | 2 - 3 | 297 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
