Assay Details
Gene Symbol: Rcbtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108380.1 | NP_001101850.1 | 4 - 5 | 817 | 63 | |
XM_008770759.1 | XP_008768981.1 | 5 - 6 | 643 | 63 | ||
GenBank mRNA | AY724518.1 | - | 3 - 4 | 463 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
