Assay Details
Gene Symbol: Rcbtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108380.1 | NP_001101850.1 | 9 - 10 | 1539 | 126 | |
XM_008770759.1 | XP_008768981.1 | 10 - 11 | 1365 | 126 | ||
GenBank mRNA | AY724518.1 | - | 8 - 9 | 1185 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P000121 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P000127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290473 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13306 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P000121 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P730048 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P000121 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P730048 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P000121 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P730048 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P162232 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 101075 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3552 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 58417 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10780823 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10780831 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10780829 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10780824 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10780830 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10780822 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10780826 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10780827 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10780828 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10780820 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10780833 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10780825 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10780832 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10780819 | - | - |
Rat Genome 230 2.0 Array | 1390433_at | - | - |
Rat Genome 230 2.0 Array | 1390433_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390433_at | - | - |
Affymetrix HT Rat Focus Array | 1390433_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1500004109.rn.1 | TC1500000586.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500004117.rn.1 | TC1500000586.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500004112.rn.1 | TC1500000586.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500004108.rn.1 | TC1500000586.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500004110.rn.1 | TC1500000586.rn.1 | RTA TC |
Rat Expression Set 230 | 1390433_PM_at | - | - |
Rat Expression Set 230 | 1390433_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10780819 | - | - |
Rat Genome U34 Set | rc_AI102429_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22406300 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370224 | - | - |
