Assay Details
Gene Symbol: Ngdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107263.1 | NP_001100733.1 | 2 - 3 | 126 | 84 | |
XM_006251966.2 | XP_006252028.1 | 2 - 3 | 319 | 84 | ||
XM_008770676.1 | XP_008768898.1 | 2 - 3 | 1111 | 84 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_006251966.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
