Assay Details
Gene Symbol: Lrp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037777.1 | NP_001032866.1 | 5 - 6 | 1818 | 88 | |
GenBank mRNA | BC100651.1 | - | 5 - 6 | 1818 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
