Assay Details
Gene Symbol: Tm9sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251990.2 | XP_006252052.1 | 5 - 6 | 1640 | 69 | |
XM_006251991.2 | XP_006252053.1 | 5 - 6 | 1656 | 69 | ||
XM_008770688.1 | XP_008768910.1 | 4 - 5 | 1857 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
