Assay Details
Gene Symbol: Myh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017239.2 | NP_058935.2 | 32 - 33 | 4994 | 74 | |
GenBank mRNA | BC070964.1 | - | 571 | 74 | ||
X15938.1 | - | 32 - 33 | 4991 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
