Assay Details
Gene Symbol: Ssbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760647.1 | XP_008758869.1 | 11 - 12 | 904 | 118 | |
XM_008760649.1 | XP_008758871.1 | 11 - 12 | 879 | 118 | ||
XM_008760651.1 | XP_008758873.1 | 10 - 11 | 813 | 118 | ||
XM_008760653.1 | XP_008758875.1 | 10 - 11 | 788 | 118 | ||
GenBank mRNA | FQ221531.1 | - | 11 - 12 | 779 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
