Assay Details
Gene Symbol: Mrps27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199192.1 | NP_001186121.1 | 8 - 9 | 725 | 117 | |
GenBank mRNA | CB576151.1 | - | 3 - 4 | 252 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
