Assay Details
Gene Symbol: Cd55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022269.2 | NP_071605.2 | 6 - 7 | 907 | 157 | |
XM_006249717.2 | XP_006249779.1 | 6 - 7 | 929 | 157 | ||
XM_006249719.2 | XP_006249781.1 | 6 - 7 | 927 | 157 | ||
GenBank mRNA | AB026900.1 | - | 6 - 7 | 914 | 157 | |
AB026901.1 | - | 6 - 7 | 851 | 157 | ||
AB026902.1 | - | 6 - 7 | 914 | 157 | ||
AB026903.1 | - | 6 - 7 | 851 | 157 | ||
AB026904.1 | - | 5 - 6 | 700 | 157 | ||
AF039583.1 | - | 6 - 7 | 848 | 157 | ||
AF039584.1 | - | 6 - 7 | 848 | 157 | ||
BC061869.1 | - | 6 - 7 | 871 | 157 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
