Assay Details
Gene Symbol: Pigr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012723.4 | NP_036855.3 | 1883 | 127 | ||
GenBank mRNA | X15741.1 | - | 5 - 6 | 1777 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
