Assay Details
Gene Symbol: Prrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250151.2 | XP_006250213.1 | 3 - 4 | 1663 | 94 | |
GenBank mRNA | AY730764.1 | - | 3 - 4 | 649 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
