Assay Details
Gene Symbol: Prrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153821.1 | NP_722543.1 | 3 - 4 | 930 | 81 | |
GenBank mRNA | S82911.1 | - | 3 - 4 | 930 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription by RNA polymerase II
positive regulation of mesenchymal cell proliferation
embryonic limb morphogenesis
inner ear morphogenesis
middle ear morphogenesis
positive regulation of smoothened signaling pathway
positive regulation of transcription by RNA polymerase II
neuron fate determination
embryonic cranial skeleton morphogenesis
embryonic skeletal system morphogenesis
artery morphogenesis
cartilage development
roof of mouth development
regulation of neuron projection regeneration
neuronal stem cell population maintenance
DNA binding
transcription coactivator activity
sequence-specific DNA binding
HMG box domain binding
negative regulation of transcription by RNA polymerase II
positive regulation of mesenchymal cell proliferation
embryonic limb morphogenesis
inner ear morphogenesis
middle ear morphogenesis
positive regulation of smoothened signaling pathway
positive regulation of transcription by RNA polymerase II
neuron fate determination
embryonic cranial skeleton morphogenesis
embryonic skeletal system morphogenesis
artery morphogenesis
cartilage development
roof of mouth development
regulation of neuron projection regeneration
neuronal stem cell population maintenance
DNA binding
transcription coactivator activity
sequence-specific DNA binding
HMG box domain binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P479890 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P093507 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737605 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919543 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539582 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38794 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P093507 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P479890 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P093507 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P479890 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P093507 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P479890 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P479890 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700537335 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48639 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92024 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 72661 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 26288 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 67638 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700537335 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10769428 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10769427 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10769424 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10769426 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10769423 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10769425 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10769429 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10769422 | - | - |
Rat Genome 230 2.0 Array | 1384234_at | - | - |
Rat Genome 230 2.0 Array | 1375485_PM_at | - | - |
Rat Genome 230 2.0 Array | 1384234_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375485_at | - | - |
Affymetrix HT Rat Focus Array | 1384234_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375485_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375485_at | - | - |
Affymetrix HT Rat Focus Array | 1384234_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1300012679.rn.1 | TC1300001786.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300012680.rn.1 | TC1300001786.rn.1 | RTA TC |
Rat Expression Set 230 | 1375485_PM_at | - | - |
Rat Expression Set 230 | 1384234_PM_at | - | - |
Rat Expression Set 230 | 1375485_at | - | - |
Rat Expression Set 230 | 1384234_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10769422 | - | - |
Rat Genome U34 Set | S82911_at | - | - |
Rat Genome U34 Set | rc_AA957064_g_at | - | - |
Rat Genome U34 Set | rc_AA957064_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368514 | - | - |
RatRef-12 Expression BeadChip | ILMN_63084 | - | - |
