Assay Details
Gene Symbol: Mybph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006249888.2 | XP_006249950.1 | 8 - 9 | 1375 | 113 | |
XM_006249889.2 | XP_006249951.1 | 8 - 9 | 1374 | 113 | ||
GenBank mRNA | BC061993.1 | - | 8 - 9 | 1304 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
