Assay Details
Gene Symbol: Ube2t | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108344.2 | NP_001101814.2 | 4 - 5 | 437 | 144 | |
GenBank mRNA | AW141189.1 | - | 4 - 5 | 437 | 144 | |
BC167012.1 | - | 4 - 5 | 398 | 144 | ||
FQ233263.1 | - | 4 - 5 | 425 | 144 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC167012.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
