Assay Details
Gene Symbol: Acbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011906.1 | NP_001011906.1 | 1 - 2 | 715 | 114 | |
XM_006250031.2 | XP_006250093.1 | 1 - 2 | 771 | 114 | ||
XM_006250032.2 | XP_006250094.1 | 1 - 2 | 779 | 114 | ||
XM_006250033.2 | XP_006250095.1 | 1 - 2 | 778 | 114 | ||
XM_006250035.2 | XP_006250097.1 | 1 - 2 | 772 | 114 | ||
XM_006250036.2 | XP_006250098.1 | 1 - 2 | 772 | 114 | ||
GenBank mRNA | BC086598.1 | - | 1 - 2 | 715 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
