Assay Details
Gene Symbol: Impa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172224.1 | NP_757378.1 | 4 - 5 | 572 | 68 | |
XM_006254856.2 | XP_006254918.1 | 4 - 5 | 3891 | 68 | ||
GenBank mRNA | AY160191.1 | - | 4 - 5 | 572 | 68 | |
BC083544.1 | - | 4 - 5 | 546 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
