Assay Details
Gene Symbol: Pggt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031082.1 | NP_112344.1 | 2 - 3 | 272 | 115 | |
XM_006254691.2 | XP_006254753.1 | 2 - 3 | 1512 | 115 | ||
XM_008772177.1 | XP_008770399.1 | 3 - 4 | 274 | 115 | ||
GenBank mRNA | L24116.1 | - | 2 - 3 | 272 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
