Assay Details
Gene Symbol: Pggt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031082.1 | NP_112344.1 | 6 - 7 | 666 | 135 | |
XM_006254691.2 | XP_006254753.1 | 6 - 7 | 1906 | 135 | ||
XM_008772177.1 | XP_008770399.1 | 7 - 8 | 668 | 135 | ||
GenBank mRNA | L24116.1 | - | 6 - 7 | 666 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
