Assay Details
Gene Symbol: Srfbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005536.1 | NP_001005536.1 | 2 - 3 | 214 | 73 | |
GenBank mRNA | BC082077.1 | - | 2 - 3 | 214 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
