Assay Details
Gene Symbol: Poli | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106137.1 | NP_001099607.1 | 6 - 7 | 995 | 149 | |
XM_006254900.2 | XP_006254962.1 | 6 - 7 | 916 | 149 | ||
XM_006254901.2 | XP_006254963.1 | 5 - 6 | 858 | 149 | ||
XM_008772110.1 | XP_008770332.1 | 3 - 4 | 850 | 149 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
