Assay Details
Gene Symbol: Smad4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019275.2 | NP_062148.1 | 6 - 7 | 957 | 83 | |
GenBank mRNA | AB010954.1 | - | 6 - 7 | 957 | 83 | |
AF056002.1 | - | 1029 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
