Assay Details
Gene Symbol: Alpk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006254887.2 | XP_006254949.1 | 6 - 7 | 6005 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
