Assay Details
Gene Symbol: Psmg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106138.2 | NP_001099608.2 | 2 - 3 | 298 | 90 | |
XM_008772117.1 | XP_008770339.1 | 2 - 3 | 327 | 90 | ||
GenBank mRNA | CB798672.1 | - | 2 - 3 | 309 | 90 | |
FQ212569.1 | - | 2 - 3 | 362 | 90 | ||
FQ230678.1 | - | 2 - 3 | 326 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
