Assay Details
Gene Symbol: Snx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106135.2 | NP_001099605.2 | 4 - 5 | 498 | 137 | |
XM_006254716.2 | XP_006254778.1 | 4 - 5 | 524 | 137 | ||
GenBank mRNA | BC166853.1 | - | 4 - 5 | 473 | 137 | |
CB557295.1 | - | 2 - 3 | 102 | 137 | ||
CO565239.1 | - | 4 - 5 | 498 | 137 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
