Assay Details
Gene Symbol: Gnal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191836.1 | NP_001178765.1 | 9 - 10 | 1174 | 88 | |
GenBank mRNA | S80376.1 | - | 9 - 10 | 1576 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
